Product Details

SNP ID
rs5775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109087635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTATTTGTGGCCTGGGCATTGT[A/G]GGCAACATCATGGTAGTCCTGGTTG
Phenotype
MIM: 188545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRHR PubMed Links

Gene Details

Gene
TRHR
Gene Name
thyrotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003301.5 212 Silent Mutation GTA,GTG V41V NP_003292.1
XM_011517263.2 212 Silent Mutation GTA,GTG V41V XP_011515565.1

View Full Product Details