Product Details

SNP ID
rs1873510
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:158491628 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATACGCTGTTCTCATGAGCTGGA[G/A]AGTATGTGGACAACTGATAGCAACT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
OR10R3P PubMed Links

Gene Details

Gene
OR10R3P
Gene Name
olfactory receptor family 10 subfamily R member 3 pseudogene
There are no transcripts associated with this gene.

View Full Product Details