Product Details
- SNP ID
-
rs952094
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
80
- Location
-
Chr.1:158614301 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGAGTCTCTTCACTCACACCTTTG[A/G]TGTCCCTGAAAGAAAAAAAAAAAAC
- Phenotype
-
MIM: 182860
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10Z1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs556396059] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10Z1
- Gene Name
- olfactory receptor family 10 subfamily Z member 1
There are no transcripts associated with this gene.
- Gene
- SPTA1
- Gene Name
- spectrin alpha, erythrocytic 1
View Full Product Details