Product Details

SNP ID
rs1056037
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:151851521 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TCACATATAATTCAAAAGAATAATA[C/G]TAAACAGTAAAATAAGAGCAGAAAA
Phenotype
MIM: 615653
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100132111 PubMed Links

Gene Details

Gene
LOC100132111
Gene Name
uncharacterized LOC100132111
There are no transcripts associated with this gene.

Gene
THEM5
Gene Name
thioesterase superfamily member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182578.3 Intron NP_872384.1
XM_011509421.1 Intron XP_011507723.1

View Full Product Details