Product Details
- SNP ID
-
rs9050
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
47
- Location
-
Chr.1:152106838 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGGAAGTTTTTATGCAGAGGTCACT[A/C]CCCCTGCACTATTCAAGGGTTATAA
- Phenotype
-
MIM: 190370
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TCHH
PubMed Links
Gene Details
- Gene
- TCHH
- Gene Name
- trichohyalin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007113.3 |
6474 |
UTR 3 |
|
|
NP_009044.2 |
View Full Product Details