Product Details

SNP ID
rs3819100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114296754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTCCCCACTAATGTGAGTCATA[C/T]AGATGGAGTCTCAGGGCACGACTGG
Phenotype
MIM: 600008
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NNMT PubMed Links

Gene Details

Gene
NNMT
Gene Name
nicotinamide N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006169.2 Intron NP_006160.1

View Full Product Details