Product Details
- SNP ID
-
rs12579103
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:15919028 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGTGTGAAATACATACACAGAGT[C/G]AATAATTATGTACATTTTAGATTTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DERA
PubMed Links
Gene Details
- Gene
- DERA
- Gene Name
- deoxyribose-phosphate aldolase
View Full Product Details