Product Details

SNP ID
rs1805723
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:8989701 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGGCAACCCAAGCCCAGAATGACTA[C/T]GGACCACAGCAAAAATGTGAGTTAA
Phenotype
MIM: 604874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRG1 PubMed Links

Gene Details

Gene
KLRG1
Gene Name
killer cell lectin like receptor G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005810.3 622 Silent Mutation TAC,TAT Y22Y NP_005801.3
XM_017018682.1 622 Silent Mutation TAC,TAT Y22Y XP_016874171.1
XM_017018683.1 622 Silent Mutation TAC,TAT Y22Y XP_016874172.1
XM_017018684.1 622 Silent Mutation TAC,TAT Y22Y XP_016874173.1
XM_017018685.1 622 Silent Mutation TAC,TAT Y22Y XP_016874174.1

View Full Product Details