Product Details

SNP ID
rs2060915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41259483 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTAAGTTGAGGATCCAGTTAGA[C/T]AGAAGCGAAAAAGTTCTGCTAGTTG
Phenotype
MIM: 191342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UCHL1 PubMed Links

Gene Details

Gene
UCHL1
Gene Name
ubiquitin C-terminal hydrolase L1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004181.4 Intron NP_004172.2
Gene
UCHL1-AS1
Gene Name
UCHL1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details