Product Details

SNP ID
rs11837125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:41197469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGAGTCAGTCTCCACAAGCAA[C/G]TATGTTAACCAGTTTCTAGCTATTT
Phenotype
MIM: 609730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDZRN4 PubMed Links

Gene Details

Gene
PDZRN4
Gene Name
PDZ domain containing ring finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164595.1 Intron NP_001158067.1
NM_013377.3 Intron NP_037509.3

View Full Product Details