Product Details

SNP ID
rs12516117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55263434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTTGCCTAAAATGTTTTTCTA[C/T]GTTTAATATGTGCTGATCAGCAAAA
Phenotype
MIM: 612720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX29 PubMed Links

Gene Details

Gene
DHX29
Gene Name
DEAH-box helicase 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019030.2 Intron NP_061903.2
XM_006714653.2 Intron XP_006714716.1
XM_017009588.1 Intron XP_016865077.1

View Full Product Details