Product Details

SNP ID
rs2839163
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:46234474 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCACACCAATTGCTAACAAACTCA[C/G]GCTTATTCCCTTTGAATTTCACCTG
Phenotype
MIM: 600909 MIM: 603294
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LSS PubMed Links
Additional Information
For this assay, SNP(s) [rs17183438] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LSS
Gene Name
lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase)
There are no transcripts associated with this gene.

Gene
MCM3AP
Gene Name
minichromosome maintenance complex component 3 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003906.4 Intron NP_003897.2
XM_005261203.4 Intron XP_005261260.1
XM_005261204.4 Intron XP_005261261.1
XM_005261205.3 Intron XP_005261262.1
Gene
MCM3AP-AS1
Gene Name
MCM3AP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details