Product Details
- SNP ID
-
rs11672
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:113977980 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGTGACAAGAAAGTTCCTATCTG[C/T]CATCTAGATCAAGAGCCTGAACTAC
- Phenotype
-
MIM: 605182
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100507747
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145101424] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100507747
- Gene Name
- uncharacterized LOC100507747
There are no transcripts associated with this gene.
- Gene
- RASA3
- Gene Name
- RAS p21 protein activator 3
View Full Product Details