Product Details

SNP ID
hCV11381449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46719750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTCGCTGCTCCAGCGGGTCCGGC[A/G]AGCCAACACCTTCTTGGAGGAGGTG
Phenotype
MIM: 176930
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F2 PubMed Links

Gene Details

Gene
F2
Gene Name
coagulation factor II, thrombin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000506.4 150 Missense Mutation CAA,CGA Q43R NP_000497.1
NM_001311257.1 150 Missense Mutation CAA,CGA Q27R NP_001298186.1

View Full Product Details