Product Details

SNP ID
rs2576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66040996 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACTGGATAAAGAGTTGCTGATAG[C/T]AGTGTCCTGGTTCTTCCCTTTACAT
Phenotype
MIM: 608234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAL3ST3 PubMed Links

Gene Details

Gene
GAL3ST3
Gene Name
galactose-3-O-sulfotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033036.2 3089 UTR 3 NP_149025.1
XM_017018519.1 3089 Intron XP_016874008.1

View Full Product Details