Product Details

SNP ID
rs1052570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66337377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAATTCCAGTCCTTGGAATCTGA[A/G]AAGCAGACACCAAGAACTGCCCTGA
Phenotype
MIM: 605517 MIM: 606259 MIM: 605965
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GAT1 PubMed Links

Gene Details

Gene
B4GAT1
Gene Name
beta-1,4-glucuronyltransferase 1
There are no transcripts associated with this gene.

Gene
BRMS1
Gene Name
breast cancer metastasis suppressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024957.1 1393 UTR 3 NP_001020128.1
NM_015399.3 1393 UTR 3 NP_056214.1
Gene
RIN1
Gene Name
Ras and Rab interactor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004292.2 1393 Intron NP_004283.2
XM_011545399.2 1393 Intron XP_011543701.1
XM_011545400.2 1393 Intron XP_011543702.1
XM_011545401.2 1393 Intron XP_011543703.1
XM_017018587.1 1393 Intron XP_016874076.1

View Full Product Details