Product Details

SNP ID
rs3167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61349135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGTGCGTCTTTGGAGAAGGCA[A/G]AAAAGCCACAGCAGCAACACTTAGG
Phenotype
MIM: 615844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYB561A3 PubMed Links

Gene Details

Gene
CYB561A3
Gene Name
cytochrome b561 family member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161452.1 2061 UTR 3 NP_001154924.1
NM_001161454.1 2061 UTR 3 NP_001154926.1
NM_001300763.1 2061 UTR 3 NP_001287692.1
NM_153611.4 2061 UTR 3 NP_705839.3
XM_011544821.1 2061 UTR 3 XP_011543123.1
XM_011544823.1 2061 UTR 3 XP_011543125.1
XM_011544824.1 2061 Intron XP_011543126.1
XM_017017345.1 2061 UTR 3 XP_016872834.1
XM_017017346.1 2061 UTR 3 XP_016872835.1
XM_017017347.1 2061 UTR 3 XP_016872836.1
Gene
TKFC
Gene Name
triokinase and FMN cyclase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015533.3 2061 Intron NP_056348.2
XM_011544912.2 2061 Intron XP_011543214.1
XM_017017517.1 2061 Intron XP_016873006.1
XM_017017518.1 2061 Intron XP_016873007.1
XM_017017519.1 2061 Intron XP_016873008.1
XM_017017520.1 2061 Intron XP_016873009.1
XM_017017521.1 2061 Intron XP_016873010.1
XM_017017522.1 2061 Intron XP_016873011.1
XM_017017523.1 2061 Intron XP_016873012.1
XM_017017524.1 2061 Intron XP_016873013.1

View Full Product Details