Product Details

SNP ID
rs2054780
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:207048267 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GGCACCTATTCCCTCATACATTCTA[C/T]GTTTTATATGCTAGAGATCTTAAAC
Phenotype
MIM: 171835 MIM: 612023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PFKFB2 PubMed Links

Gene Details

Gene
PFKFB2
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2
There are no transcripts associated with this gene.

Gene
YOD1
Gene Name
YOD1 deubiquitinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276320.1 1847 UTR 3 NP_001263249.1
NM_018566.3 1847 UTR 3 NP_061036.3

View Full Product Details