Product Details

SNP ID
rs3800342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41191794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGTGAGAGCGTGGGCTCTGCG[C/G]GGTCCCAGGACAGGAGGGAGGAGCA
Phenotype
MIM: 609715
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TREML2 PubMed Links

Gene Details

Gene
TREML2
Gene Name
triggering receptor expressed on myeloid cells like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024807.3 1503 UTR 3 NP_079083.2
XM_011514917.2 1503 UTR 3 XP_011513219.1

View Full Product Details