Product Details

SNP ID
rs1927522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:26758699 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTGAACGATGTCATTGTTTCACG[A/G]ATGCTAAGTGCTCCTGTGGCTTGAG
Phenotype
MIM: 600752
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR12 PubMed Links

Gene Details

Gene
GPR12
Gene Name
G protein-coupled receptor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005288.3 1351 UTR 3 NP_005279.1
XM_005266360.3 1351 UTR 3 XP_005266417.1

View Full Product Details