Product Details

SNP ID
rs1546585
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:43144609 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGATGCGTGGGCCGCCAGCTTCAG[A/T]CCTGCTTTTCTTTCTGATGGCCCTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929767 PubMed Links

Gene Details

Gene
LOC101929767
Gene Name
uncharacterized LOC101929767
There are no transcripts associated with this gene.

Gene
NBR2
Gene Name
neighbor of BRCA1 gene 2 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details