Product Details
- SNP ID
-
rs3729617
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:43083086 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGTTGGCCGGTTCAGACTCTAGT[A/G]TTGGAGCCAAGAGCAGTTTGGGGCC
- Phenotype
-
MIM: 601890
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KLC4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116363087] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLC4
- Gene Name
- kinesin light chain 4
There are no transcripts associated with this gene.
- Gene
- PTK7
- Gene Name
- protein tyrosine kinase 7 (inactive)
View Full Product Details