Product Details

SNP ID
rs10256915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:137228476 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTCACTGCTCAATGGAAAACTAC[A/G]AGAGCTCTGCTTAGCCCCTCATTTC
Phenotype
MIM: 162095
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTN PubMed Links

Gene Details

Gene
PTN
Gene Name
pleiotrophin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321386.1 Intron NP_001308315.1
NM_001321387.1 Intron NP_001308316.1
NM_002825.5 Intron NP_002816.1
XM_017012473.1 Intron XP_016867962.1

View Full Product Details