Product Details
- SNP ID
-
rs10249257
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:123661753 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGCAGAGGATTATTAACTTTTTT[A/T]ATTATAAACTTATGTCACTTGCCAT
- Phenotype
-
MIM: 608006
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LMOD2
PubMed Links
Gene Details
- Gene
- LMOD2
- Gene Name
- leiomodin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207163.1 |
|
Intron |
|
|
NP_997046.1 |
View Full Product Details