Product Details

SNP ID
rs5468
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:72054432 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGCCAGGTTTCTTGGGATTTGTAA[G/T]AGAACATCACAAGAAAATCAAGTGT
Phenotype
MIM: 140100 MIM: 140210
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HP PubMed Links

Gene Details

Gene
HP
Gene Name
haptoglobin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126102.2 Intron NP_001119574.1
NM_001318138.1 Intron NP_001305067.1
NM_005143.4 Intron NP_005134.1
Gene
HPR
Gene Name
haptoglobin-related protein
There are no transcripts associated with this gene.

Gene
TXNL4B
Gene Name
thioredoxin like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142317.1 Intron NP_001135789.1
NM_001142318.1 Intron NP_001135790.1
NM_001324354.1 Intron NP_001311283.1
NM_001324355.1 Intron NP_001311284.1
NM_017853.2 Intron NP_060323.1
XM_017023377.1 Intron XP_016878866.1

View Full Product Details