Product Details

SNP ID
rs878983812
Assay Type
DME
NCBI dbSNP Submissions
1
Location
Chr.1:109669568 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TGGCCAAACTCTGCTATGACCCAGA[A/T]TTTGTAAGTCCCCCCACCCCACTCC
Phenotype
MIM: 138380 MIM: 138333
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GSTM2 PubMed Links

Gene Details

Gene
GSTM2
Gene Name
glutathione S-transferase mu 2 (muscle)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000848.3 451 Missense Mutation GAA,GAT E119D NP_000839.1
NM_001142368.1 451 Missense Mutation GAA,GAT E119D NP_001135840.1
Gene
GSTM4
Gene Name
glutathione S-transferase mu 4
There are no transcripts associated with this gene.

View Full Product Details