Product Details
- SNP ID
-
rs1935662
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
38
- Location
-
Chr.1:205714495 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGCACCACTGTGAATTTTGTGAA[G/T]TTGAAAAAAAAAAGTGTAATTTATG
- Phenotype
-
MIM: 611912
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NUCKS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144482900] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NUCKS1
- Gene Name
- nuclear casein kinase and cyclin dependent kinase substrate 1
View Full Product Details