Product Details

SNP ID
rs2013809
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:3278261 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGCAGCTGCCACGTGGCTGTAGGC[A/G]GTGATGATGAGAACCAAAGGTGTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR3A2 PubMed Links

Gene Details

Gene
OR3A2
Gene Name
olfactory receptor family 3 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002551.3 714 Silent Mutation ACC,ACT T225T NP_002542.3
XM_017024697.1 714 Silent Mutation ACC,ACT T227T XP_016880186.1
XM_017024698.1 714 Silent Mutation ACC,ACT T227T XP_016880187.1

View Full Product Details