Product Details

SNP ID
rs1920144
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:171141168 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTAAAAGCCGTGAGAAAACAAGTAT[C/T]AGAATAACTATAAGTGACTATATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMO6P PubMed Links

Gene Details

Gene
FMO6P
Gene Name
flavin containing monooxygenase 6 pseudogene
There are no transcripts associated with this gene.

View Full Product Details