Product Details

SNP ID
rs812936
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:5844638 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACAGGGATGTGGAAAGGCCATGTCC[A/G]TAGCAGGATCAGGAGGGTGGGGCGG
Phenotype
MIM: 111100 MIM: 136836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUT3 PubMed Links

Gene Details

Gene
FUT3
Gene Name
fucosyltransferase 3 (Lewis blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000149.3 840 Missense Mutation CGG,TGG R68W NP_000140.1
NM_001097639.1 840 Missense Mutation CGG,TGG R68W NP_001091108.1
NM_001097640.1 840 Missense Mutation CGG,TGG R68W NP_001091109.1
NM_001097641.1 840 Missense Mutation CGG,TGG R68W NP_001091110.1
XM_011527865.1 840 Missense Mutation CGG,TGG R68W XP_011526167.1
XM_011527866.1 840 Missense Mutation CGG,TGG R68W XP_011526168.1
XM_011527867.1 840 Missense Mutation CGG,TGG R68W XP_011526169.1
Gene
FUT6
Gene Name
fucosyltransferase 6
There are no transcripts associated with this gene.

Gene
LOC101928844
Gene Name
uncharacterized LOC101928844
There are no transcripts associated with this gene.

View Full Product Details