Product Details

SNP ID
rs887806
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:39866607 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TATTGAGATAAATACAAATCCAGAG[A/G]CTAAACCGCAGAGACCTGGATAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FCGBP PubMed Links

Gene Details

Gene
FCGBP
Gene Name
Fc fragment of IgG binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003890.2 Intron NP_003881.2

View Full Product Details