Product Details
- SNP ID
-
rs887803
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39866420 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTATCCTGGGATTTTCTAGCTACA[G/T]GGACTAATAAATTCAATTTTGCTTA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FCGBP
PubMed Links
Gene Details
- Gene
- FCGBP
- Gene Name
- Fc fragment of IgG binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003890.2 |
|
Intron |
|
|
NP_003881.2 |
View Full Product Details