Product Details
- SNP ID
-
hCV11472188
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:231712461 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCAGGTGAGGAAGAGGATGGAG[A/G]TGAAGATGAGGAAGCTGAGTCAGCT
- Phenotype
-
MIM: 188390
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR1244-1
PubMed Links
Gene Details
- Gene
- MIR1244-1
- Gene Name
- microRNA 1244-1
There are no transcripts associated with this gene.
- Gene
- PTMA
- Gene Name
- prothymosin, alpha
View Full Product Details