Product Details

SNP ID
rs2036878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231061020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCAATGCTGTTAGTCCAGGGA[A/C]CACACTGAAAACCATTGAATTAGGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PSMD1 PubMed Links

Gene Details

Gene
PSMD1
Gene Name
proteasome 26S subunit, non-ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191037.1 Intron NP_001177966.1
NM_002807.3 Intron NP_002798.2
XM_017004517.1 Intron XP_016860006.1

View Full Product Details