Product Details

SNP ID
rs1894529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37643701 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAAGGCCTAGGAGGCAGCCCGGG[C/T]AGTCACAGCCATACGACCATGGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927051 PubMed Links

Gene Details

Gene
LOC101927051
Gene Name
uncharacterized LOC101927051
There are no transcripts associated with this gene.

Gene
SH3BP1
Gene Name
SH3 domain binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018957.3 642 Silent Mutation GGC,GGT G177G NP_061830.3

View Full Product Details