Product Details
- SNP ID
-
rs11635
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:32388064 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGATTCCAGCATCATGGCAGGTATT[C/T]ACCACATCTGTCACATTCTTATAGG
- Phenotype
-
MIM: 613901
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC339666
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs576737790] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC339666
- Gene Name
- uncharacterized LOC339666
There are no transcripts associated with this gene.
- Gene
- RTCB
- Gene Name
- RNA 2',3'-cyclic phosphate and 5'-OH ligase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014306.4 |
1577 |
Silent Mutation |
GTA,GTG |
V482V |
NP_055121.1 |
View Full Product Details