Product Details
- SNP ID
-
rs1920739
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:90968520 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGGATTTATTTATGTATTAAAAAT[A/G]CAGAAGTTGAAGACAATGTAAGCTA
- Phenotype
-
MIM: 601657
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPYC
PubMed Links
Gene Details
- Gene
- EPYC
- Gene Name
- epiphycan
View Full Product Details