Product Details
- SNP ID
-
rs10698
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:131524009 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGCTTTCCAGAGACAGCCACCAC[G/T]CAGGAGGGAGGATCACCCCAGGCAA
- Phenotype
-
MIM: 607423
MIM: 609328
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
POMT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111722104] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POMT1
- Gene Name
- protein O-mannosyltransferase 1
- Gene
- UCK1
- Gene Name
- uridine-cytidine kinase 1
View Full Product Details