Product Details
- SNP ID
-
hCV11489715
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:110911160 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTCACGTCAGGGGGGAAGGCGG[C/T]GAACATCTGGTCAACCTGCAATGAG
- Phenotype
-
MIM: 160781
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC63
PubMed Links
Gene Details
- Gene
- CCDC63
- Gene Name
- coiled-coil domain containing 63
There are no transcripts associated with this gene.
- Gene
- MYL2
- Gene Name
- myosin light chain 2
There are no transcripts associated with this gene.
View Full Product Details