Product Details

SNP ID
rs3744183
Assay Type
Functionally Tested
NCBI dbSNP Submissions
25
Location
Chr.17:79079585 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
ACTTCAGCCACCACACCGTCACCAT[C/T]CCCCCAGTGGGCTGGACCAACACTG
Phenotype
MIM: 611898 MIM: 616999
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ENGASE PubMed Links

Gene Details

Gene
ENGASE
Gene Name
endo-beta-N-acetylglucosaminidase
There are no transcripts associated with this gene.

Gene
RBFOX3
Gene Name
RNA binding protein, fox-1 homolog 3
There are no transcripts associated with this gene.

View Full Product Details