Product Details

SNP ID
rs8107444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52800274 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAACCTTGCCACATTCATTACAC[A/G]TGTATGGTTTCTCTCCAGTATGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF28 PubMed Links

Gene Details

Gene
ZNF28
Gene Name
zinc finger protein 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006969.3 1758 Intron NP_008900.3
XM_006723354.3 1758 Intron XP_006723417.1
XM_011527262.2 1758 Missense Mutation XP_011525564.1

View Full Product Details