Product Details

SNP ID
rs7582569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:109131499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTCTTCTGTTCTGATAAATTAT[A/G]TGATATTCGGAGCCCCGCTTCTAGT
Phenotype
MIM: 601181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4265 PubMed Links

Gene Details

Gene
MIR4265
Gene Name
microRNA 4265
There are no transcripts associated with this gene.

Gene
RANBP2
Gene Name
RAN binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006267.4 Intron NP_006258.3
XM_005264002.2 Intron XP_005264059.1
XM_005264003.2 Intron XP_005264060.1
XM_005264004.2 Intron XP_005264061.1
XM_005264005.4 Intron XP_005264062.1
XM_005264007.2 Intron XP_005264064.1
XM_011511575.2 Intron XP_011509877.1
XM_011511576.2 Intron XP_011509878.1
XM_011511578.2 Intron XP_011509880.1
XM_017004623.1 Intron XP_016860112.1
XM_017004624.1 Intron XP_016860113.1
XM_017004625.1 Intron XP_016860114.1
Gene
SH3RF3
Gene Name
SH3 domain containing ring finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099289.2 Intron NP_001092759.1
XM_011511109.2 Intron XP_011509411.1
XM_011511110.2 Intron XP_011509412.1
Gene
SH3RF3-AS1
Gene Name
SH3RF3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details