Product Details
- SNP ID
-
rs6571653
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:21090399 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCCACCCTCCTACGCCCGCCGC[C/G]CCTTCCACCTCTGGTCCGCCTGGGG
- Phenotype
-
MIM: 610018
MIM: 605036
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141238761,rs200238223] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF40
- Gene Name
- Rho guanine nucleotide exchange factor 40
- Gene
- TMEM253
- Gene Name
- transmembrane protein 253
There are no transcripts associated with this gene.
- Gene
- ZNF219
- Gene Name
- zinc finger protein 219
View Full Product Details