Product Details

SNP ID
rs12590397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24038614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTTTAGTCTCAAAGAAAGTAT[A/G]TAAGTCAGTGTCACAGTTAGTAGTG
Phenotype
MIM: 615195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS4L1 PubMed Links

Gene Details

Gene
DHRS4L1
Gene Name
dehydrogenase/reductase 4 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277864.1 Intron NP_001264793.1
XM_006720246.3 Intron XP_006720309.1

View Full Product Details