Product Details

SNP ID
rs12583132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19268641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATGGAGGAAATGATACTTGTCA[G/T]TCTGTGATTATGAAATACATGTATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD26P3 PubMed Links

Gene Details

Gene
ANKRD26P3
Gene Name
ankyrin repeat domain 26 pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details