Product Details

SNP ID
rs12616061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240887100 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCCTCGCCTCCCAGAGTCCTGGT[C/T]TTCTGGAAGATGCTCTGCATGGCAG
Phenotype
MIM: 604285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGXT PubMed Links

Gene Details

Gene
AGXT
Gene Name
alanine-glyoxylate aminotransferase
There are no transcripts associated with this gene.

Gene
C2orf54
Gene Name
chromosome 2 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085437.2 1473 Silent Mutation AAA,AAG K438K NP_001078906.2
NM_001282921.1 1473 Silent Mutation AAA,AAG K289K NP_001269850.1
NM_024861.3 1473 Silent Mutation AAA,AAG K270K NP_079137.3
XM_011511877.1 1473 Silent Mutation AAA,AAG K438K XP_011510179.1

View Full Product Details