Product Details

SNP ID
rs4846199
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:10223903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATATTTAGGTTTAGGTTGTTTTG[A/G]GAAAGTAATTAAATATAAACTTCTA
Phenotype
MIM: 605995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF1B PubMed Links

Gene Details

Gene
KIF1B
Gene Name
kinesin family member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015074.3 Intron NP_055889.2
NM_183416.3 Intron NP_904325.2
Gene
MIR1273D
Gene Name
microRNA 1273d
There are no transcripts associated with this gene.

View Full Product Details