Product Details

SNP ID
rs11739096
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180503106 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAATGCCGGTGGTGCAGTGGGCT[C/G]TGCTGCTTTACATGTGTCTCATTTA
Phenotype
MIM: 608951
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CNOT6 PubMed Links

Gene Details

Gene
CNOT6
Gene Name
CCR4-NOT transcription complex subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303241.1 Intron NP_001290170.1
XM_005265953.1 Intron XP_005266010.1
XM_011534605.2 Intron XP_011532907.1
XM_011534606.1 Intron XP_011532908.1
XM_011534607.1 Intron XP_011532909.1
XM_017009670.1 Intron XP_016865159.1
XM_017009671.1 Intron XP_016865160.1
XM_017009672.1 Intron XP_016865161.1

View Full Product Details