Product Details

SNP ID
rs2374215
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:113902035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTCACCACGTTTTCTAAAGGAGC[A/G]AGAGAAATAAACATCTCCACTCTCT
Phenotype
MIM: 610010
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARSJ PubMed Links

Gene Details

Gene
ARSJ
Gene Name
arylsulfatase family member J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024590.3 3522 UTR 3 NP_078866.3
XM_005263212.4 3522 UTR 3 XP_005263269.1
XM_011532238.2 3522 UTR 3 XP_011530540.1
XM_011532239.2 3522 UTR 3 XP_011530541.1
XM_017008592.1 3522 UTR 3 XP_016864081.1
XM_017008593.1 3522 UTR 3 XP_016864082.1
XM_017008594.1 3522 UTR 3 XP_016864083.1
XM_017008595.1 3522 UTR 3 XP_016864084.1
XM_017008596.1 3522 UTR 3 XP_016864085.1
XM_017008597.1 3522 UTR 3 XP_016864086.1
XM_017008598.1 3522 UTR 3 XP_016864087.1
XM_017008599.1 3522 UTR 3 XP_016864088.1
XM_017008600.1 3522 UTR 3 XP_016864089.1

View Full Product Details