Product Details

SNP ID
rs12650984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:90142596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAATACAGGCAAATGCACAATACA[A/G]CAGAGGGATATCGTTTTATTTATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCSER1 PubMed Links

Gene Details

Gene
CCSER1
Gene Name
coiled-coil serine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145065.1 Intron NP_001138537.1
NM_207491.2 Intron NP_997374.1
XM_011531936.1 Intron XP_011530238.1
XM_011531937.2 Intron XP_011530239.1
XM_011531938.2 Intron XP_011530240.1
XM_011531939.2 Intron XP_011530241.1
XM_011531941.2 Intron XP_011530243.1
XM_011531942.2 Intron XP_011530244.1
XM_011531943.2 Intron XP_011530245.1
XM_011531945.1 Intron XP_011530247.1
XM_011531946.2 Intron XP_011530248.1
XM_011531947.2 Intron XP_011530249.1
XM_011531948.1 Intron XP_011530250.1
XM_011531949.2 Intron XP_011530251.1
XM_011531950.2 Intron XP_011530252.1
XM_011531951.2 Intron XP_011530253.1
XM_011531955.2 Intron XP_011530257.1
XM_011531956.2 Intron XP_011530258.1
XM_011531957.2 Intron XP_011530259.1
XM_011531958.2 Intron XP_011530260.1
XM_017008194.1 Intron XP_016863683.1
XM_017008195.1 Intron XP_016863684.1
XM_017008196.1 Intron XP_016863685.1
XM_017008197.1 Intron XP_016863686.1
XM_017008198.1 Intron XP_016863687.1
XM_017008199.1 Intron XP_016863688.1
XM_017008200.1 Intron XP_016863689.1
XM_017008201.1 Intron XP_016863690.1
XM_017008202.1 Intron XP_016863691.1

View Full Product Details